Find Courses
  • Search by date
What's New
Training Login
Training Cart
  • You do not have any classes in your cart.
Contact Us
Digital PCR – Absolute Quantitation (4:00)
Share on:
Who should attend?
Anyone who would like to learn how digital PCR can be used in absolute quantification.
Course Description
This video discusses the use of digital PCR in absolute quantification assays. Content includes:
  • how classifying partitioned reactions as either positive or negative allows for absolute quantification without the need for a reference standard.
  • considerations to keep in mind when considering absolute quantification by digital PCR.
Tell a colleague about this course. Copy the following URL:
https://learn.thermofisher.com/courses/view/id/371
CBD   Carlsbad, CA   FRD   Frederick, MD   REG   Regional   WEB   Web-Based Training
Price
$0.00  
Click thumbnails to watch videos
(Roll over thumbnails to view descriptions)